Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Water testing kit, MINIKIT

Water testing kit, MINIKIT

Sulphite (low range), 2 - 50 mg/l Na2SO3, AF 434, 1 KIT
N° de catalogue LOVI414340
Fournisseur:   Lovibond
Water testing kit, MINIKIT
N° de catalogue LOVI414340
Fournisseur:  Lovibond
Numéro CAS:  
Water testing kit, MINIKIT
Water testing kit, MINIKIT

Sulphite (low range), 2 - 50 mg/l Na2SO3, AF 434, 1 KIT

Numéro CAS:  
Prix
63,90 €
Connexion Pour la disponibilité
1 * 1 KIT
Product Details & Documents

MINIKIT provides quick and simple tests for the analysis of boiler water, cooling water, drinking and industrial water.

  • MINIKITS are based on titrimetric methods
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with a minimum guaranteed shelf life of five years

Tablet count method: In this method the liquid titration solution and indicator are replaced by AQUALYTIC® tablet reagents. A specific number of tablets is added to a defined sample volume until a chemically induced colour change takes place. The concentration of the parameter being measured is calculated from the number of tablets required. The measuring range can be expanded by varying the sample volume.

Yes/No test: This method tells whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.

Turbidity method: A two-section calibrated test tube is filled with the water sample and a reagent tablet added. The reagent creates a level of turbidity that is proportional to the concentration of the parameter being measured. The inner tube, which has a black dot on its base, is lowered until the dot is obscured by the turbidity. The result is read off from the water level in the inner tube.

Documents

Spécifications

Spécifications

  • Cdt:
    1 KIT
  • Désignation:
    Sulphite (low range)
  • Type:
    AF 434
  • Plage de mesure:
    2 - 50 mg/l Na2SO<sub>3</sub>

Spécifications

Accessories

Informations sur le produit

  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • LOVI385130
    Dilution/shaker tube with lid, plastic, graduated, 100 ml
    Connexion Pour la disponibilité
    1 * 1 ST - 9,90 €

Informations sur le produit

  • N° de catalogue
    Désignation
    Disponibilité
    Prix
  • LOVI385132
    Dilution/shaker tube with lid, plastic, graduated, 30 ml
    Connexion Pour la disponibilité
    1 * 1 ST - 9,90 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.