Stoppers, with drip tip, MBL®
29/32, With pointed bottom
N° de catalogue 217-0749
Fournisseur: SciLabware
Aller à :
Product Details & Documents
Ideally suited for use with volumetric flasks etc. Lower closed end of stopper features a 'drip-tip' to ensure adhered liquid is returned to the flask. Flat head allows stopper to be stood on end, thus preventing contamination of the ground surface.
- Hexagonal head prevents the stopper from rolling and provides a good grip
- Hollow-blown to reduce weight
Spécifications
- RN:29/32
- Type:With pointed bottom
- Seq:07