Dropping funnels, cylindrical, Quickfit®
Dropping funnels, cylindrical, Quickfit®
100 ml, 19/26 NS, 19/26 NS
N° de catalogue 532-0076
Fournisseur:   SciLabware
Dropping funnels, cylindrical, Quickfit®
N° de catalogue 532-0076
Fournisseur:  SciLabware
Dropping funnels, cylindrical, Quickfit®
Dropping funnels, cylindrical, Quickfit®
100 ml, 19/26 NS, 19/26 NS
Prix
87,80 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Funnel stem with ground-glass cone for flasks and assemblies. Stopcock has interchangeable glass key.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
 : Complies with ISO 4800
Spécifications
  • Rodage mâle:
    19/26 NS
  • Rodage femelle:
    19/26 NS
  • Capacité:
    100 ml
  • Seq:
    02