Rabbit serum
Rabbit serum
Sérum de lapin, Filtré de manière stérile, Frozen
N° de catalogue S2500-100
Fournisseur:   Biowest
Rabbit serum
N° de catalogue S2500-100
Fournisseur:  Biowest
Rabbit serum
Rabbit serum
Sérum de lapin, Filtré de manière stérile, Frozen
Prix
17,90 €
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents
Rabbit serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Rabbit serum is used as a blocking agent in immunoassays and controls.
Spécifications
  • Format du sérum:
    Frozen
  • Stérilité:
    Filtré de manière stérile
  • Désignation:
    Sérum de lapin