DEPC
Flacon Verre, 100 mL
Catalog # SERA18835.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- CAS Number:1609-47-8
- Formula:O(COOC₂H₅)₂
- MDL Number:MFCD00009106
- Molecular Weight:162.14 g/mol
- Merck Index:13,08088
- Boiling Pt:93 °C (1013 hPa)
- Density:1,123 g/cm³ (20 °C)
- Flash Pt:69 °C
- Storage Temperature:Réfrigérateur
- EINECS:216-542-8
Specifications
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:100 mL



