Agarose
Glass bottle for solids, 250 g
N° de catalogue SERA11384.03
Marketsource
Fournisseur: Serva
Aller à :
Product Details & Documents
- Numéro CAS:9012-36-6
- Numéro MDL:MFCD00081294
- Point de fusion:60…90 °C
- Température de stockage:Température ambiante
- EINECS:232-731-8
Spécifications
Spécifications
- Type de conditionnement:Glass bottle for solids
- Cdt:250 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



