Base materials
Peptone from meat (pancreatic digest), Flacon verre pour solides, 1 kg
N° de catalogue SERA48619.02
Marketsource
Fournisseur: Serva
Aller à :
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
- Suitable as nutrient for laboratory media and industrial fermentation
Spécifications
Spécifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:1 kg
- Désignation:Peptone from meat (pancreatic digest)