Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Reference standards VOA

Reference standards VOA

Bromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1
Catalog # REST30252
Supplier:   Restek
Reference standards VOA
Catalog # REST30252
Supplier:  Restek
CAS Number:  
Reference standards VOA

Bromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule, 1

CAS Number:  
Price
54,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Cdt:
    1
  • Désignation:
    Bromoform, 2000 µg/ml, purge and trap grade methanol, 1 ml/ampule
  • Cat. No.:
    REST30252
  • N ° CAS.:
    75-25-2
  • Formula:
    CHBr₃
  • Désignation:
    Bromoform

Specifications