Magnetic surface temperature probes
Magnetic surface probe up to 250 °C (for TFN 520), Lemosa connection, 1 m (Silicone cable), TPN 900, −50...+250 °C
N° de catalogue EBRO1341-0640
Fournisseur: EBRO
Aller à :
Product Details & Documents
These probes are used for surface temperature measurement on an extended period of ferrous containing compounds, e.g. heating plates, heating tubes, pumps, flushing tools, motors and turbines.
Spécifications
- Plage de mesure:−50...+250 °C
- Désignation:Magnetic surface probe up to 250 °C (for TFN 520), Lemosa connection, 1 m (Silicone cable)
- Seq:3
- Type:TPN 900
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



