Screw caps for micro tubes
Screw caps for micro tubes, violet
N° de catalogue SARS65.716.008
Marketsource
Fournisseur: SARSTEDT
Aller à :
Product Details & Documents
PP, coloured.
- Excellent chemical resistance
- Excellent temperature resistance
Screw cap with O-ring seal.
Spécifications
- Couleur:violet
- Désignation:Screw caps for micro tubes
- Seq:10