Empty solid phase extraction columns, CHROMABOND®
Reservoir columns, PP, with one adapter for 15, 45, 70 ml CHROMABOND® PP columns, 70 ml
Catalog # MANA730389
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Empty and reservoir columns with accessories for solid phase extraction.
- Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
- Reservoir columns from PP for application of medium-size samples
- Accessories for SPE columns (filter elements, adapters)
Specifications
- Capacité:70 ml
- Désignation:Reservoir columns, PP, with one adapter for 15, 45, 70 ml CHROMABOND® PP columns
- Seq:18
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- MANA732019PE frits for LV PP columns 15 ml, for 100 mg adsorbent weightLog in for availability1 * 250 ST - 132,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
- MANA732020PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weightLog in for availability1 * 250 ST - 185,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- MANA730105Adapters (PVDF) for glass columns (3 and 6 ml)Log in for availability1 * 10 ST - 200,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- MANA730385Adapters (PE) for PP columns (15, 45 and 70 ml)Log in for availability1 * 10 ST - 125,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice