Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Extraction thimbles, borosilicate glass

43×123 mm, 2,0 mm, MN 649, for dust analysis in hot exhaust gases
  MANA649131
Extraction thimbles, borosilicate glass
  MANA649131
Extraction thimbles, borosilicate glass
Extraction thimbles, borosilicate glass

43×123 mm, 2,0 mm, MN 649, for dust analysis in hot exhaust gases

 
338,00 €
   
1 * 25 ST
 

Made from glass micro fibers without binder.

  • Pollution monitoring, air sampling
  • High temperature filtration (up to 500 °C)
  • For solvents incompatible with cellulose thimbles (for example strong acids or bases)

Extraction thimbles for high temperatures and aggressive solvents

 
  • Ø int.×h:
    43×123 mm
  • Type:
    MN 649, for dust analysis in hot exhaust gases
  • Épaisseur:
    2,0 mm