Extraction thimbles, borosilicate glass
15×50 mm, 1,0 mm, MN 649, for dust analysis in hot exhaust gases
Made from glass micro fibers without binder.
- Pollution monitoring, air sampling
- High temperature filtration (up to 500 °C)
- For solvents incompatible with cellulose thimbles (for example strong acids or bases)
Extraction thimbles for high temperatures and aggressive solvents
- Ø int.×h:15×50 mm
- Type:MN 649, for dust analysis in hot exhaust gases
- Épaisseur:1,0 mm



