Extraction thimbles
33×205 mm, 1,5 mm
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
- Fat determination in food and feed
- Determination of binder content in asphalt
- Analysis of soil contaminants
- Determination of additives in rubber and elastomers
Specifications
- Ø int.×h:33×205 mm
- Seq:26
- Épaisseur:1,5 mm