Extraction thimbles
28× 80 mm, 1,5 mm
Fournisseur: MACHEREY-NAGEL
Aller à :
Product Details & Documents
Extraction thimbles, made of pure cellulose, standard quality. With or without lid available.
- Fat determination in food and feed
- Determination of binder content in asphalt
- Analysis of soil contaminants
- Determination of additives in rubber and elastomers
Spécifications
- Ø int.×h:28× 80 mm
- Seq:12
- Épaisseur:1,5 mm