Sleeves for ground joints, rigid
Sleeves for ground joints, rigid
45/50
N° de catalogue COWI020.145
Fournisseur:   COWIE
Sleeves for ground joints, rigid
N° de catalogue COWI020.145
Fournisseur:  COWIE
Sleeves for ground joints, rigid
Sleeves for ground joints, rigid
45/50
Prix
28,40 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Made of PTFE, grease-free, high vacuum full length rigid sleeves for standard glass joints, with substantial shoulder for ease of handling and repeated use.
  • Rigid sleeves and adapters are inert
  • Usable to 280˚C high vacuum
  • The leakage rate is less
  • High vacuum performance is obtained by transverse sealing rings
Spécifications
  • Pour rodage:
    45/50
  • Seq:
    07