Bottle pourers
Bottle pourer, PTFE, blue, for GL 38 screw neck bottles
N° de catalogue COWI028.038
Fournisseur: COWIE
Aller à :
Product Details & Documents
Manufactured in pure, FDA approved, PTFE.
- Totally chemically resistant and inert
- To fit GL45, 38, 32 and 30 screw neck bottles
- State cap colour required, blue, red or green
Permits direct pouring in a safe and consistent manner.
Spécifications
- Type:for GL 38 screw neck bottles
- Désignation:Bottle pourer, PTFE, blue