Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
24/40, 55 mm
Catalog # COWI010.124
Supplier:   COWIE
Stoppers, flat type, PTFE
Catalog # COWI010.124
Supplier:  COWIE
Stoppers, flat type, PTFE
Stoppers, flat type, PTFE
24/40, 55 mm
Price
18,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
PTFE. Stopper with flat head fit standard ground glass joints.
  • Trapezoidal rings give high vacuum performance and reduce seizure
Specifications
  • RN:
    24/40
  • Hauteur:
    55 mm