Printers
Printer, AP24XS-40-V, 58 mm, 113,5×96×66 mm, 391 g
  LANC39010005
Printers
  LANC39010005
Printers
Printer, AP24XS-40-V, 58 mm, 113,5×96×66 mm, 391 g
1 * 1 ST
  • Largeur d'impression (maximale):
    58 mm
  • Poids:
    391 g
  • Désignation:
    Printer, AP24XS-40-V
  • Seq:
    02
  • l×p×h:
    113,5×96×66 mm