Restore™ Western Blot Stripping Buffer
Restore™ Western Blot Stripping Buffer
Restore™ Western Blot Stripping Buffer, trial pack, 30 mL
N° de catalogue PIER21062
Fournisseur:   Thermo Fisher Scientific
Restore™ Western Blot Stripping Buffer
N° de catalogue PIER21062
Numéro CAS:  
Restore™ Western Blot Stripping Buffer
Restore™ Western Blot Stripping Buffer
Restore™ Western Blot Stripping Buffer, trial pack, 30 mL
Numéro CAS:  
Prix
64,50 €
Connexion Pour la disponibilité
1 * 30 mL
Product Details & Documents
Although optimising assay conditions is the best way to achieve optimum results, re-performing the gel electrophoresis process to test each new primary antibody or antibody concentration is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
  • Saves time - no need to re-run gels
  • Saves precious sample
  • Provides efficient removal
  • Gentle formulation ensures target protein is not damaged after stripping and re-probing
  • Odour-free
Spécifications
Spécifications
  • Cdt:
    30 mL
  • Désignation:
    Restore™ Western Blot Stripping Buffer, trial pack
Spécifications
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.