Restore™ Western Blot Stripping Buffer
Restore™ Western Blot Stripping Buffer, trial pack, 30 mL
N° de catalogue PIER21062
Fournisseur: Thermo Fisher Scientific
Restore™ Western Blot Stripping Buffer
N° de catalogue PIER21062
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Aller à :
Product Details & Documents
Although optimising assay conditions is the best way to achieve optimum results, re-performing the gel electrophoresis process to test each new primary antibody or antibody concentration is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
- Saves time - no need to re-run gels
- Saves precious sample
- Provides efficient removal
- Gentle formulation ensures target protein is not damaged after stripping and re-probing
- Odour-free
Spécifications
Spécifications
- Cdt:30 mL
- Désignation:Restore™ Western Blot Stripping Buffer, trial pack
Spécifications
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.