Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Glass sample tubes for rack R-12 polyvap, round bottom (Ø 48 mm), height 170 mm, for working volume up to 120 ml
N° de catalogue BUCH040907
Fournisseur:   BUCHI
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
N° de catalogue BUCH040907
Fournisseur:  BUCHI
Accessories for parallel synthesis, Syncore® Polyvap, Syncore® Analyst
Glass sample tubes for rack R-12 polyvap, round bottom (Ø 48 mm), height 170 mm, for working volume up to 120 ml
Prix
516,00 €
Connexion Pour la disponibilité
1 * 12 ST
Product Details & Documents
Spécifications
  • Désignation:
    Glass sample tubes for rack R-12 polyvap, round bottom (Ø 48 mm), height 170 mm, for working volume up to 120 ml