Bouchons compte-gouttes, WHEATON®
20 - 410, rouge
Catalogus # WHEAW242522-A
Leverancier: DWK Life Sciences
Ga naar:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specificaties
- Filetage:20 - 410
- Seq:47
- Couleur bouchon:rouge