Dispenser bottles, Wheaton®
125 ml, PE-BD, 130 mm, 43 mm, 20-410
Catalog # WHEA211195
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These translucent dispensing bottles are excellent for mild and strong buffers with a spout to dispense the contents and then could be depressed to seal the bottle.
- Very flexible with high impact strength
- Good chemical resistance
Specifications
- Ø ext.:43 mm
- Filetage:20-410
- Capacité:125 ml
- Hauteur:130 mm
- Matière:PE-BD
- Seq:02



