Snap caps for sample bottles, WHEATON®
45 mm, 60/120 ml
N° de catalogue WHEA242619
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
Natural low density polyethylene.
- All sizes have tabs for easy removal
- Good chemical resistance
- Good temperature resistance
For WHEATON snap cap sample bottles.
Spécifications
- Taille du bouchon:45 mm
- Pour flacons:60/120 ml