Bouchons compte-gouttes, WHEATON®
13 - 425, orange
N° de catalogue 215-4030
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Spécifications
- Filetage:13 - 425
- Seq:17
- Couleur bouchon:orange