Poids de calibration, classe F2
2 g, 0,4 ± mg, Poids à gorge de préhension sans certificat
Catalog # SARBYCW324-00
Supplier: Sartorius Balances
Jump to:
Product Details & Documents
Weights are made of steel and are supplied in plastic or wooden boxes.
Specifications
- Poids:2 g
- Tolérance:0,4 ± mg
- Type:Poids à gorge de préhension sans certificat
Accessories
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW32Pinces avec revêtement de silicone
pointes, 160 mm, pour 1 g - 200 gLog in for availability1 * 1 ST - 36,40 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW05Cloche en verre avec plaque de support, pour 20 kgLog in for availability1 * 1 ST - 1 660,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW31Pinces avec revêtement de silicone
pointes, 115 mm, pour 1 mg - 5 gLog in for availability1 * 1 ST - 26,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW02Cloche en verre avec plaque de support, pour 100 g - 1 kg (2 kg)Log in for availability1 * 1 ST - 936,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW33Pinces avec revêtement de silicone
pointes, 230 mm, pour 1 g - 1 kgLog in for availability1 * 1 ST - 57,20 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW04Cloche en verre avec plaque de support, pour 10 kgLog in for availability1 * 1 ST - 1 460,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW01Cloche en verre avec plaque de support, pour 1 mg - 50 g (100 g ou 200 g)Log in for availability1 * 1 ST - 832,00 €
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW20GLPlastic screw box for 1 - 20 g weights with closed-pore foam inletLog in for availability1 * 1 ST - 12,40 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW100GLPlastic screw-top boxes for 100 g weights with closed-pore foam inletLog in for availability1 * 1 ST - 15,60 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW50GLPlastic screw box for 50 g weights with closed-pore foam inletLog in for availability1 * 1 ST - 15,60 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW2000GLPlastic screw box for 1 - 2000 g weights with closed-pore foam inletLog in for availability1 * 1 ST - 33,30 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW500GLPlastic screw box for 1 - 500 g weights with closed-pore foam inletLog in for availability1 * 1 ST - 20,80 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW5000GLPlastic screw box for 1 - 5000 g weights with closed-pore foam inletLog in for availability1 * 1 ST - 46,80 €
Product Information
- Catalog #DésignationAvailabilityPrice
- SARBYAW01GLPlastic screw box for 100 mg weights with closed-pore foam inletLog in for availability1 * 1 ST - 12,40 €
Product Information
- Catalog #DésignationAvailabilityPrice