Flasks, round bottom, Quickfit® ground joints
5000 ml, 29/32 NS, 270 mm
N° de catalogue 201-8546
Fournisseur: SciLabware
Aller à :
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certifications: ISO 1773, ISO 4797.
Documents
Spécifications
- Rodage femelle:29/32 NS
- Capacité:5000 ml
- Hauteur:270 mm



