Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Swan neck gouge, double curved, -, 25 mm, 290 mm
  AESCND118R
 :   Aesculap
Gouges, Aesculap®
  AESCND118R
 :  Aesculap
Gouges, Aesculap®

Swan neck gouge, double curved, -, 25 mm, 290 mm

 
 
   
1 * 1 ST
 
 
  • Type:
    -
  • Largeur:
    25 mm
  • Désignation:
    Swan neck gouge, double curved
  • Longueur:
    290 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.