Dilator set, Aesculap®
Dilator, 15 mm
N° de catalogue AESCEM115R
Fournisseur: Aesculap
Aller à :
Product Details & Documents
Complete set of uterine dilators.
Delivery information: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).
Spécifications
- Désignation:Dilator, 15 mm
- Seq:16