Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Anoscopes, Bensaude, Aesculap®

Anoscopes, Bensaude, Aesculap®

Anoscope, Bensaude, 55 mm, 20 mm
Catalog # AESCEA863C
Supplier:   Aesculap
Anoscopes, Bensaude, Aesculap®
Catalog # AESCEA863C
Supplier:  Aesculap
Anoscopes, Bensaude, Aesculap®

Anoscope, Bensaude, 55 mm, 20 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Anoscopes for abdominal surgery.

Specifications
  • Ø:
    20 mm
  • Désignation:
    Anoscope, Bensaude
  • Longueur:
    55 mm
  • Seq:
    02