Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Forceps, micro, jeweller pattern, Aesculap®

Forceps, micro, jeweller pattern, Aesculap®

Straight, very fine (0,4 mm), with teeth 1:2, 135 mm
Catalog # AESCBD345R
Supplier:   Aesculap
Forceps, micro, jeweller pattern, Aesculap®
Catalog # AESCBD345R
Supplier:  Aesculap
Forceps, micro, jeweller pattern, Aesculap®

Straight, very fine (0,4 mm), with teeth 1:2, 135 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    Straight, very fine (0,4 mm), with teeth 1:2
  • Longueur:
    135 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.