Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Forceps, micro, jeweller pattern, Aesculap®

Straight, very fine (0,3 mm), 135 mm
N° de catalogue AESCBD335R
Fournisseur:   Aesculap
Forceps, micro, jeweller pattern, Aesculap®
N° de catalogue AESCBD335R
Fournisseur:  Aesculap
Forceps, micro, jeweller pattern, Aesculap®

Straight, very fine (0,3 mm), 135 mm

Prix
51,30 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Type:
    Straight, very fine (0,3 mm)
  • Longueur:
    135 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.