Béchers, forme basse
250 ml, , 85 mm, 65 mm
N° de catalogue BOCH8511
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
Stainless steel, 18/10.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Spécifications
- Ø ext.:65 mm
- Capacité:250 ml
- Hauteur:85 mm