Béchers, forme basse
2000 ml, , 180 mm, 120 mm
N° de catalogue BOCH8524
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
Stainless steel, 18/10.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Spécifications
- Ø ext.:120 mm
- Capacité:2000 ml
- Hauteur:180 mm