Béchers, forme basse
5000 ml, , 210 mm, 180 mm
N° de catalogue BOCH8526
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
Stainless steel, 18/10.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Spécifications
- Ø ext.:180 mm
- Capacité:5000 ml
- Hauteur:210 mm