Hexane (mélange d'isomères), TECHNICAL
Fut métallique, 25 L
Supplier: VWR Chemicals
Hexane (mélange d'isomères), TECHNICAL
Catalog # 24574.460
Supplier: VWR Chemicals
CAS Number: 110-54-3
Jump to:
Product Details & Documents
Synonyms: Hexanes, Isohexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD02179311
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:68…70 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0,66 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Température ambiante
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
P20_VWRC24574.323_SPEC_FR.pdf P20_VWRC24574.367_SPEC_FR.pdf P20_VWRC24574.460_SPEC_FR.pdf P20_VWRC24574.551_SPEC_FR.pdf spec_VWRC24574.298
Product specification
More
VWR Chemicals Big Packs and Bulks VWR® for rotary evaporation
Product Literature
Specifications
Specification Test Results
- Boiling point:63 - 69 °C
- Evaporation residue:Max. 20 ppm
Specifications
- Type de conditionnement:Fut métallique
- Cdt:25 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



