Volumetric overflow flasks, without stopper
Volumetric overflow flasks, without stopper
43,5 ml, Overflow flask
N° de catalogue NEUB0372-10-0435
Volumetric overflow flasks, without stopper
N° de catalogue NEUB0372-10-0435
Volumetric overflow flasks, without stopper
43,5 ml, Overflow flask
Prix
26,40 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These volumetric overflow flasks are made of borosilicate glass 3.3.
  • Calibrated to deliver (TD, Ex)
  • Cut and ground exactly at the volume-mark
  • Without stopper
Specially designed for water analysis.
Spécifications
  • Capacité:
    43,5 ml
  • Désignation:
    Overflow flask
  • Seq:
    01