Spore strips, Attest™
Spore strips, Attest™
Spores/bandelette de bacillus stearothermophilus, stérilisation à la vapeur
N° de catalogue MMMEM1262
Fournisseur:   3M MEDICAL
Spore strips, Attest™
N° de catalogue MMMEM1262
Fournisseur:  3M MEDICAL
Spore strips, Attest™
Spore strips, Attest™
Spores/bandelette de bacillus stearothermophilus, stérilisation à la vapeur
Prix
338,00 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
3M™ Attest™ Biological Monitoring Systems are true biological indicators, in self-contained indicator capsules. They enable checks on the biological reliability and performance of each steriliser without laboratory facilities and laboratory trained personnel.
  • Self-contained, reducing the risk of false positives due to contamination
  • Colour change gives clear unambiguous, visual indication of PASS or FAIL
  • Time saving and results in 24 to 48 hours which require no interpretation
Spécifications
  • Désignation:
    Spores/bandelette de bacillus stearothermophilus, stérilisation à la vapeur