Wire baskets, autoclavable, Poxygrid®
280×150×90 mm
N° de catalogue BELAH167650000
Fournisseur: Bel-Art Products, a Part of SP
Wire baskets, autoclavable, Poxygrid®
N° de catalogue BELAH167650000
Fournisseur: Bel-Art Products, a Part of SP
Aller à :
Product Details & Documents
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
- Resistant to rust, corrosion, solvents, acids and alkalis
- Withstands a temperature range of −223 to +121 °C
Spécifications
- l×p×h:280×150×90 mm