Hypodermic needles
21, 25,4 mm
  POPR4050
Hypodermic needles
  POPR4050
Hypodermic needles
Hypodermic needles
21, 25,4 mm
1 170,00 €
1 * 12 Pce
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
  • Jauge:
    21
  • Longueur:
    25,4 mm
  • Seq:
    15