Hypodermic needles
22, 25,4 mm
POPR4043
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
- Jauge:22
- Longueur:25,4 mm
- Seq:17