Hypodermic needles
25, 9,5 mm
N° de catalogue POPR4017
Fournisseur: CADENCE SCIENCE
Aller à :
Product Details & Documents
Hypodermic needles are precision made and the points are machine ground to achieve uniform sharpness. The needle cannula are fabricated of type 304 stainless steel tubing. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure they are burrs and debris free.
Spécifications
- Jauge:25
- Longueur:9,5 mm
- Seq:25