Acétonitrile ≥99%, TECHNICAL
Fut métallique, 25 L
Supplier: VWR Chemicals
Acétonitrile ≥99%, TECHNICAL
Catalog # 20070.462
Supplier: VWR Chemicals
CAS Number: 75-05-8
Jump to:
Product Details & Documents
Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS Number:75-05-8
- Formula:H₃CCN
- MDL Number:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:13,00071
- Boiling Pt:81,6 °C (1013 hPa)
- Melting Pt:–45,7 °C
- Density:0,782 g/cm³ (20 °C)
- Flash Pt:2 °C (coupe fermée)
- Storage Temperature:Température ambiante
- EINECS:200-835-2
- UN:1648
- ADR:3,II
- REACH:01-2119471307-38
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125)
Specifications
Specification Test Results
- Assay:Min. 99 %
- Indice de réfraction (20°C):1.343 - 1.345
Specifications
- Type de conditionnement:Fut métallique
- Cdt:25 L
- Cat. No.:20070.462
- Certification environnementale:Chem21 Solvent Selection Guide recommended by default
- Environmentally Preferable:Safer Human and Environmental Health



