Micro-pipettes, à usage unique, étalonnées
25 µl, 2 × blanc
DRUM2-000-025
: DRUMMOND
These precision calibrated micro pipettes are available in sizes ranging from 1 to 200 µl. Ideal for a variety of uses in liquid transfer work.
- Fire polished
- Accuracy of ±0,5%
- Colour coded micro pipettes with sizes imprinted
- Anticoagulant treatment for clinical applications (special limited sizes)
: 250 colour coded micro pipettes, plus one 15" aspirator tube, are supplied in a conveniently designed, easy to store cylinder.
- Code couleur:2 × blanc
- Capacité:25 µl