Micro-pipettes, à usage unique, étalonnées
5 µl, blanc
  DRUM2-000-005
 :   DRUMMOND
Micro-pipettes, à usage unique, étalonnées
  DRUM2-000-005
 :  DRUMMOND
Micro-pipettes, à usage unique, étalonnées
5 µl, blanc
102,00 €
1 * 250 ST
These precision calibrated micro pipettes are available in sizes ranging from 1 to 200 µl. Ideal for a variety of uses in liquid transfer work.
  • Fire polished
  • Accuracy of ±0,5%
  • Colour coded micro pipettes with sizes imprinted
  • Anticoagulant treatment for clinical applications (special limited sizes)
 : 250 colour coded micro pipettes, plus one 15" aspirator tube, are supplied in a conveniently designed, easy to store cylinder.
  • Code couleur:
    blanc
  • Capacité:
    5 µl
Accessories
  •  
    Désignation
  • DRUM2-000-000
    Aspirator tube assembly, 38 cm
    1 * 1 ST - 7,20 €