BOD bottles
BOD bottle, with glass stopper, 300 ml
N° de catalogue HACH62100
Fournisseur: Hach
Aller à :
Product Details & Documents
These are standard bottles, made of borosilicate glass, for BOD analaysis.
- Flared lip and ground-glass stopper to prevent air entrapment and for water seal
- Serialised bottles have silk-screened, sequential numbers and include a bar code
- Frosted marking area
Livraison: Stoppers are included.
Spécifications
- Capacité:300 ml
- Désignation:BOD bottle, with glass stopper
- Seq:01