Eau, TECHNICAL, déminéralisée
Cubitainer, 10 L
Eau, TECHNICAL, déminéralisée
23597.405
: 7732-18-5
: Hydrogen oxide
- :7732-18-5
- :H₂O
- :MFCD00011332
- :18.02 g/mol
- :13,10098
- :100 °C (1013 hPa)
- :0 °C
- :1 g/cm³ (4 °C)
- :Température ambiante
- :231-791-2
Documents
10L Cubitainer UN (ZPACK13260004) 10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 200L Plastic drums for liquids UN (ZPACK13950512) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Packaging Information
P20_VWRC23597.405_SPEC_FR.pdf P20_VWRC23597.410_SPEC_FR.pdf P20_VWRC23597.440_SPEC_FR.pdf P20_VWRC23597.553_SPEC_FR.pdf spec_VWRC23597.405
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
- Conductivity (20°C) (at production):Max. 1.5 µS/cm
- Type de conditionnement:Cubitainer
- Cdt:10 L
- Cat. No.:23597.405
- Certification environnementale:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



