Bouchons compte-gouttes, WHEATON®
15 - 415, blanc
N° de catalogue 215-4019
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Spécifications
- Filetage:15 - 415
- Seq:32
- Couleur bouchon:blanc