Flacons à tare
45 ml, 35×70 mm
  611-0099
Flacons à tare
  611-0099
Flacons à tare
Flacons à tare
45 ml, 35×70 mm
21,90 €
1 * 1 ST
Made of borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
With interchangeable lid and frosted label.
  • Ø×h:
    35×70 mm
  • Capacité:
    45 ml
  • Seq:
    04