Cables for Screw Cap Electrodes
Cables for Screw Cap Electrodes
Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument sockets, 1 m
N° de catalogue HACHA94L114
Fournisseur:   Hach
Cables for Screw Cap Electrodes
N° de catalogue HACHA94L114
Fournisseur:  Hach
Cables for Screw Cap Electrodes
Cables for Screw Cap Electrodes
Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument sockets, 1 m
Prix
105,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spécifications
  • Désignation:
    Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument sockets
  • Longueur:
    1 m
  • Seq:
    02